Anti-ZP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
21279-1-AP-150UL
10097-398EA 524.48 USD
10097-398
Anti-ZP3 Rabbit Polyclonal Antibody
Antibodies
Zona pellucida (ZP) is an extracellular matrix that surrounds the oocyte and early embryo, which mediates species-specific sperm binding, induction of the acrosome reaction and prevents post-fertilization polyspermy. Mammalian ZP consists of three to four glycoproteins, called ZP1, ZP2, ZP3, ZP4. ZP3 (zona pellucida glycoprotein 3, also known as zona pellucida sperm-binding protein 3) is essential for sperm binding and zona matrix formation.

IHC: Human breast cancer Tissue, 1:20-1:200; Western Blot: Multi-Cells, 1:200-1:2000

Type: Primary
Antigen: ZP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR