Anti-RB1CC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
21269-1-AP-150UL
10081-260EA 402 USD
10081-260
Anti-RB1CC1 Rabbit Polyclonal Antibody
Antibodies
RB1CC1, also named as RBICC, is implicated in the regulation of RB1 expression and functions as a DNA-binding transcription factor. It is a potent regulator of the RB1 pathway and a mediator that plays a crucial role in muscular differentiation. Its expression is, thus, a prerequisite for myogenic differentiation. Involved in autophagy. RB1CC1 is required for autophagosome formation. It is probably involved in the tumorigenesis of breast cancer. RB1CC1 is frequently mutated in breast cancer and shows characteristics of a classical tumor suppressor gene. This antibody is a rabbit polyclonal antibody raised against residues near the N terminus of human RB1CC1.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: RB1CC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR