Anti-CTR9 Rabbit Polyclonal Antibody

Supplier: Proteintech
21264-1-AP-150UL
10085-326EA 524.48 USD
10085-326
Anti-CTR9 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Raji Cells, 1:500-1:5000; IHC: Human breast cancer, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: CTR9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR