Anti-ER Rabbit Polyclonal Antibody

Supplier: Proteintech
21244-1-AP-150UL
10086-388EA 537.7 USD
10086-388
Anti-ER Rabbit Polyclonal Antibody
Antibodies
The estrogen receptor (ESR, ER) is a ligand-activated transcription factor composed of several domains important for hormone binding, DNA binding, and activation of transcription. ESR1, also known as ESR or NR3A1, belongs to the nuclear hormone receptor family and NR3 subfamily. It is a nuclear hormone receptor. The steroid hormones and their receptors are involved in the regulation of eukaryotic gene expression and affect cellular proliferation and differentiation in target tissues. ESR1 can activate the transcriptional activity of TFF1.

Western Blot: MCF-7 Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:10-1:100

Type: Primary
Antigen: ER
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR