Anti-SCO2 Rabbit Polyclonal Antibody

Supplier: Proteintech
21223-1-AP-150UL
10094-218EA 402 USD
10094-218
Anti-SCO2 Rabbit Polyclonal Antibody
Antibodies
Human SCO2 is a mitochondrial membrane-bound protein involved in copper supply for the assembly of cytochrome c oxidase in eukaryotes.It is involved in the maintenance of cell copper homeostasis or, limited to HSco2, as a downstream mediator of the Warburg effect, as its expression is regulated by p53.This protein belongs to the SCO1/2 family and defects in SCO2 are the cause of fatal infantile cardioencephalomyopathy with cytochrome c oxidase deficiency (FIC).

Western Blot: HEK-293 Cells, 1:200-1:2000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: SCO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR