Anti-CPLX4 Rabbit Polyclonal Antibody

Supplier: Proteintech
21222-1-AP-150UL
10085-116EA 402 USD
10085-116
Anti-CPLX4 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Y79 Cells, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200; IP:Y79 Cells, 1:500-1:5000

Type: Primary
Antigen: CPLX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR