Anti-AAMP Rabbit Polyclonal Antibody

Supplier: Proteintech
21220-1-AP-150UL
10082-278EA 402 USD
10082-278
Anti-AAMP Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse lung Tissue, 1:1000-1:10000; IHC: Human Placenta, 1:20-1:200

Type: Primary
Antigen: AAMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR