Anti-ATOH1 Rabbit Polyclonal Antibody

Supplier: Proteintech
21215-1-AP-150UL
10083-266EA 537.7 USD
10083-266
Anti-ATOH1 Rabbit Polyclonal Antibody
Antibodies
ATOH1, also named as Protein atonal homolog 1 or BHLHA14, is a 364 amino acid protein, which contains 1 bHLH (basic helix-loop-helix) domain. ATOH1,as a transcriptional regulator in the nucleus activates E box-dependent transcription in collaboration with TCF3/E47, but the activity is completely antagonized by the negative regulator of neurogenesis HES1. ATOH1 plays a role in the differentiation of subsets of neural cells by activating E box-dependent transcription.

Western Blot:Neuro-2a Cells, 1:200-1:2000

Type: Primary
Antigen: ATOH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR