Anti-PLOD2 Rabbit Polyclonal Antibody

Supplier: Proteintech
21214-1-AP-150UL
10092-480EA 537.7 USD
10092-480
Anti-PLOD2 Rabbit Polyclonal Antibody
Antibodies
PLOD2,also named as LH2, forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. It is a potential novel prognostic factor for HCC patients following surgery. Among the PLOD genes, PLOD2 contributes to cancer prognosis and angiogenesis. Several authors have reported that PLOD2 expression might provide prognostic information about malignant tumours such as glioblastoma. PLOD2 expression is a useful biomarker for the effects of antiangiogenic treatment for malignancy. . It has 2 isoforms produced by alternative splicing and seven glycosylation sites.This antibody is specific to PLOD2.

Western Blot:DU 145 Cells, 1:200-1:2000; IHC: Human Liver cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP: Mouse Testis Tissue, 1:200-1:2000

Type: Primary
Antigen: PLOD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR