Anti-TET2 Rabbit Polyclonal Antibody

Supplier: Proteintech
21207-1-AP-150UL
10095-796EA 537.7 USD
10095-796
Anti-TET2 Rabbit Polyclonal Antibody
Antibodies
TET2 is an enzyme which converts 5-methylcytosine (5-mC) to 5-hydroxymethylcytosine (5-hmC) in DNA and is important for normal myelopoiesis. TET2 mutations are frequently observed in myeloid malignancies, including myeloproliferative neoplasms. TET2 is broadly expressed in a variety of tissues. In healthy tissues, TET2 expression was shown to be elevated in hematopoietic cells with highest expression in granulocytes. Three isoforms of TET2 exists due to the alternative splicing. 220-250 kDa bands represent the full-length isoform; 130-133 kDa bands represent the nuclear isoforms of TET2. This antibody recognizes all three isoforms of TET2.

Western Blot: HL-60 Cells, 1:200-1:2000; IP: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: TET2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR