Anti-PDE6A Rabbit Polyclonal Antibody

Supplier: Proteintech
21200-1-AP-150UL
10092-008EA 524.48 USD
10092-008
Anti-PDE6A Rabbit Polyclonal Antibody
Antibodies
PDE6A(Rod cGMP-specific 3',5'-cyclic phosphodiesterase subunit alpha) is also named as GMP-PDE alpha, PDE V-B1, PDEA and belongs to the cyclic nucleotide phosphodiesterase family. PDE6A contains an open reading frame capable of coding for a polypeptide of 859 amino acids and about 100 kDa. It is a subunit of a key phototransduction enzyme which participates in processes of transmission and amplification of the visual signal. The expression of PDE6 alpha in retina is essential for normal expression of PDE6 beta and PDE6 gama.

Western Blot: Human Brain Tissue, 1:200-1:2000; IF:retinal Tissue, N/A; IHC: Human ovary tumor Tissue, 1:20-1:200

Type: Primary
Antigen: PDE6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR