Anti-GOLGA3 Rabbit Polyclonal Antibody

Supplier: Proteintech
21193-1-AP-150UL
10087-266EA 402 USD
10087-266
Anti-GOLGA3 Rabbit Polyclonal Antibody
Antibodies
Western Blot:L02 Cells, 1:1000-1:10000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: GOLGA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR