Anti-TAOK2 Rabbit Polyclonal Antibody

Supplier: Proteintech
21188-1-AP-150UL
10095-602EA 402 USD
10095-602
Anti-TAOK2 Rabbit Polyclonal Antibody
Antibodies
TAOK2(Serine/threonine-protein kinase TAO2) is also named as KIAA0881, MAP3K17, PSK, PSK1 and belongs to the protein kinase superfamily. TAOK2 is known to modulate gene transcription via its activation of several MAP kinase pathways, including c-Jun amino-terminal kinase (JNK), and has been implicated in the modulation of actin and microtubule dynamics. The time course of TAOK2 expression is consistent with a role for it in neuronal differentiation..

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: TAOK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR