Anti-MYLK2 Rabbit Polyclonal Antibody

Supplier: Proteintech
21173-1-AP-150UL
10090-728EA 537.7 USD
10090-728
Anti-MYLK2 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:500-1:5000

Type: Primary
Antigen: MYLK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR