Anti-MKL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
21166-1-AP-150UL
10090-250EA 402 USD
10090-250
Anti-MKL1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:1000-1:10000; IHC: Human tonsillitis Tissue, 1:20-1:200

Type: Primary
Antigen: MKL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR