Anti-LILRA4 Rabbit Polyclonal Antibody

Supplier: Proteintech
21153-1-AP-150UL
10089-412EA 402 USD
10089-412
Anti-LILRA4 Rabbit Polyclonal Antibody
Antibodies
Western Blot:L02 Cells, 1:500-1:5000

Type: Primary
Antigen: LILRA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR