Anti-SLITRK6 Rabbit Polyclonal Antibody

Supplier: Proteintech
21150-1-AP-150UL
10094-822EA 402 USD
10094-822
Anti-SLITRK6 Rabbit Polyclonal Antibody
Antibodies
  • Western Blot:A549 Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100; FC: HepG2 Cells, N/A

Type: Primary
Antigen: SLITRK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR