Anti-SPACA3 Rabbit Polyclonal Antibody

Supplier: Proteintech
21137-1-AP-150UL
10095-038EA 402 USD
10095-038
Anti-SPACA3 Rabbit Polyclonal Antibody
Antibodies
SPACA3, also named as LYC3, LYZL3, SLLP1, SPRASA, CT54 and ALLP-17, belongs to the glycosyl hydrolase 22 family. It is a sperm surface membrane protein that may be involved in sperm-egg plasma membrane adhesion and fusion during fertilization. It could be a potential receptor for the egg oligosaccharide residue N-acetylglucosamine, which is present in the extracellular matrix over the egg plasma membrane. SPACA3 has a Dimer form.

Western Blot: HEK-293 Cells, 1:200-1:1000

Type: Primary
Antigen: SPACA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR