Anti-CLDN18 Rabbit Polyclonal Antibody

Supplier: Proteintech
21126-1-AP-150UL
10084-828EA 402 USD
10084-828
Anti-CLDN18 Rabbit Polyclonal Antibody
Antibodies
Claudin 18 (CLDN18) belongs to the large claudin family of proteins, which are tetraspan transmembrane proteins of tight junctions . Claudins exhibit specific patterns of expression in different tissues . CLDN18 is specifically expressed in the stomach and lung. CLDN18 has two alternatively spliced variants, CLDN18.1 and CLDN18.2. CLDN18.2 is a highly selective gastric lineage marker that determines the gastric phenotype in a neoplastic condition, whereas CLDN18.1 is lung specific . Altered CLDN18 expression has been reported in various human malignancies, including gastric cancer, lung adenocarcinoma, and pancreatic neoplasm .

Western Blot: Mouse Kidney Tissue, 1:200-1:2000; IHC: Human Stomach Tissue, 1:20-1:200; IP: Mouse Stomach Tissue, 1:200-1:2000

Type: Primary
Antigen: CLDN18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR