Anti-CHCHD4 Rabbit Polyclonal Antibody

Supplier: Proteintech
21090-1-AP-150UL
10084-428EA 537.7 USD
10084-428
Anti-CHCHD4 Rabbit Polyclonal Antibody
Antibodies
Western Blot:rat Brain Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: CHCHD4
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR