Anti-TRIM54 Rabbit Polyclonal Antibody

Supplier: Proteintech
21074-1-AP-150UL
10096-354EA 402 USD
10096-354
Anti-TRIM54 Rabbit Polyclonal Antibody
Antibodies
TRIM54(Tripartite motif-containing protein 54) is also names as MURF, MURF3, RNF30.The expression of TRIM54 is required for skeletal myoblast differentiation and myotube fusion.It is expressed in heart and skeletal muscle specifically .It has two isforms with the molecular weight of 40KDa and 45KDa.It can be dimerizated and hetero-dimerizated to form homooligomer and heterooligomer.

Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human Heart, 1:20-1:200

Type: Primary
Antigen: TRIM54
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR