Anti-SIM2 Rabbit Polyclonal Antibody

Supplier: Proteintech
21069-1-AP-150UL
10094-656EA 524.48 USD
10094-656
Anti-SIM2 Rabbit Polyclonal Antibody
Antibodies
SIM2, also named as single-minded homolog 2, is a 667 amino acid protein, which contains 1 bHLH (basic helix-loop-helix) domain and one PAS-associated C-terminal domain. SIM2 localizes in the nucleus and has tow isoforms. Isoform SIM2 is a 73 kDa protein and Isoform SIM2S is 63 kDa protein. SIM2 as a transcription factor may be a master gene of CNS development in cooperation with Arnt. It may have pleiotropic effects in the tissues expressed during development.

Western Blot:COLO 320 Cells, 1:500-1:5000

Type: Primary
Antigen: SIM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR