Anti-OXA1L Rabbit Polyclonal Antibody

Supplier: Proteintech
21055-1-AP-150UL
10091-650EA 537.7 USD
10091-650
Anti-OXA1L Rabbit Polyclonal Antibody
Antibodies
Oxa1 is a member of the conserved Oxa1/YidC/Alb3 protein family and is required for cytochrome c oxidase assembly. The Oxa1 protein was initially identified in yeast proteins involved in the biogenesis of the mitochondrial respiratory chain complexes. It locates in the inner membrane of mitochondria and may be involved in mediating the insertion of proteins from the matrix into the inner membrane.

Western Blot: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: OXA1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR