Anti-NR1I3 Rabbit Polyclonal Antibody

Supplier: Proteintech
21042-1-AP-150UL
10091-270EA 402 USD
10091-270
Anti-NR1I3 Rabbit Polyclonal Antibody
Antibodies
The nuclear hormone receptor superfamily is a large group of related transcription factors which includes members that bind a diverse array of ligands, including steroids, retinoic acid, thyroid hormone, and vitamin D. Nuclear hormone receptors contain a DNA-binding domain (DBD) and a ligand-binding domain . Nuclear receptor subfamily 1 group I member 3 (NR1I3; CAR), binds and transactivates the retinoic acid response elements that control expression of the retinoic acid receptor beta 2 and alcohol dehydrogenase 3 genes. It is expressed primarily in liver and regulates the expression of genes involved in xenobiotic metabolism as well as hormone, energy, and lipid homeostasis .

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: NR1I3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR