Anti-Willin Rabbit Polyclonal Antibody

Supplier: Proteintech
21039-1-AP-150UL
10096-602EA 402 USD
10096-602
Anti-Willin Rabbit Polyclonal Antibody
Antibodies
Willin, also known as FRMD6 or human Expanded (hEx), is a member of the FERM-domain family of cytoskeletal crosslinkers. It is a potent inhibitor of cell proliferation, transformation and modulator of drug sensitivity, suggesting it possesses tumor suppressor properties. Willin is widely expressed in normal tissues. Although willin is found in the cytoplasm and membrane, it localizes to the nuclei of almost all tumors.This antibody was raised against the C-terminal region of human FRMD6 protein and is expected to recognize the endogenous FRMD6.

IHC: Human Liver cancer Tissue, 1:20-1:200; Western Blot:Transfected HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: Willin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR