Anti-CRYBB3 Rabbit Polyclonal Antibody

Supplier: Proteintech
21009-1-AP-150UL
10085-222EA 402 USD
10085-222
Anti-CRYBB3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Eye Tissue, 1:1000-1:10000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: CRYBB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR