Anti-BRIP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20983-1-AP-150UL
10081-982EA 402 USD
10081-982
Anti-BRIP1 Rabbit Polyclonal Antibody
Antibodies
BRIP1, also named as BACH1 and FANCJ, belongs to the DEAD box helicase family and DEAH subfamily. It is a DNA-dependent ATPase and 5' to 3' DNA helicase required for the maintenance of chromosomal stability. It is involved in the repair of DNA double-strand breaks by homologous recombination in a manner that depends on its association with BRCA1. Defects in BRIP1 are a cause of susceptibility to breast cancer (BC). Defects in BRIP1 are the cause of Fanconi anemia complementation group J (FANCJ). The antibody is specific to BRIP1.

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: BRIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR