Anti-NR2C2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20981-1-AP-150UL
10091-274EA 402 USD
10091-274
Anti-NR2C2 Rabbit Polyclonal Antibody
Antibodies
NR2C2, also named as TAK1, TR4 and TR2R1, belongs to the nuclear hormone receptor family and NR2 subfamily. NR2C2 is an orphan nuclear receptor. It may regulate gene expression during the late phase of spermatogenesis. The antibody is specific to NR2C2.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: NR2C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR