Anti-INSC Rabbit Polyclonal Antibody

Supplier: Proteintech
20973-1-AP-150UL
10088-928EA 402 USD
10088-928
Anti-INSC Rabbit Polyclonal Antibody
Antibodies
INSC may function as an adapter linking the Par3 complex to the GPSM1/GPSM2 complex. It is involved in spindle orientation during mitosis it may regulate cell proliferation and differentiation in the developing nervous system. INSC may play a role in the asymmetric division of fibroblasts and participate in the process of stratification of the squamous epithelium. The antibody can recognize isoform1, 2,4,5 of INSC.

Western Blot: Mouse Liver Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200; IF: HeLa Cells, 1:10-1:100

Type: Primary
Antigen: INSC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR