Anti-CRHR1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20967-1-AP-150UL
10084-454EA 402 USD
10084-454
Anti-CRHR1 Rabbit Polyclonal Antibody
Antibodies
Corticotropin-releasing hormone (CRH, also known as Corticotropin-releasing factor, CRF) is a 41 amino acid peptide hormone and a key regulator of the stress response. CRH and the related urocortin peptides (UCN, UCN2 and UCN3) mediate their actions through two mammalian receptor subtypes, CRHR1 and CRHR2 . CRHR1 (also known as CRFR1) exhibits high affinity towards CRH and UCN but low affinity towards UCN2 and no affinity towards UCN3 . The activity of this receptor is mediated by G proteins which activate adenylyl cyclase. Widely distributed through out the brain, CRHR1 critically controls behavioral adaptation to stress and is causally linked to emotional disorders .

Western Blot:SH-SY5Y Cells, 1:200-1:1000

Type: Primary
Antigen: CRHR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR