Anti-KIAA0391 Rabbit Polyclonal Antibody

Supplier: Proteintech
20959-1-AP-150UL
10089-188EA 402 USD
10089-188
Anti-KIAA0391 Rabbit Polyclonal Antibody
Antibodies
MRPP3 (Mitochondrial ribonuclease P protein 3), originally known as KIAA0391, has been identified as a member of human mitochondrial RNase P (mtRNase P), the endonuclease responsible for tRNA 5' end maturation by removing extra nucleotides from tRNA precursors at their 5' termini. Recently Daniel et al. reported that Rnase P and other mitochondrial RNA processing enzymes are localized near mt DNA nucleoids where they can initiate process of primary transcripts. (24703694)

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: KIAA0391
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR