Anti-DPCD Rabbit Polyclonal Antibody

Supplier: Proteintech
20936-1-AP-150UL
10086-086EA 402 USD
10086-086
Anti-DPCD Rabbit Polyclonal Antibody
Antibodies
DPCD (for deleted in a mouse model of primary ciliary dyskinesia), is predicted to code for a protein of 23 kDa which remains a novel candidate gene for PCD (Primary ciliary dyskinesia).

Western Blot: Human Testis Tissue, 1:200-1:1000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: DPCD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR