Anti-PHIP Rabbit Polyclonal Antibody

Supplier: Proteintech
20933-1-AP-150UL
10091-850EA 524.48 USD
10091-850
Anti-PHIP Rabbit Polyclonal Antibody
Antibodies
Western Blot:A375 Cells, 1:1000-1:10000

Type: Primary
Antigen: PHIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR