Anti-CD62E Rabbit Polyclonal Antibody

Supplier: Proteintech
20894-1-AP-150UL
10094-280EA 524.34 USD
10094-280
Anti-CD62E Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:200-1:2000

Type: Primary
Antigen: CD62E
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR