Anti-PRKCE Rabbit Polyclonal Antibody

Supplier: Proteintech
20877-1-AP-150UL
10092-380EA 402 USD
10092-380
Anti-PRKCE Rabbit Polyclonal Antibody
Antibodies
Western Blot:SH-SY5Y Cells, 1:500-1:5000; IHC: Human thyroid cancer Tissue, 1:20-1:200

Type: Primary
Antigen: PRKCE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR