Anti-VMAT2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20873-1-AP-150UL
10096-988EA 524.48 USD
10096-988
Anti-VMAT2 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:200-1:2000

Type: Primary
Antigen: VMAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR