Anti-RHBDD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20869-1-AP-150UL
10093-638EA 402 USD
10093-638
Anti-RHBDD1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Testis Tissue, 1:500-1:5000

Type: Primary
Antigen: RHBDD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR