Anti-THAP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20853-1-AP-150UL
10095-438EA 402 USD
10095-438
Anti-THAP2 Rabbit Polyclonal Antibody
Antibodies
THAP2, also named as THAP domain-containing protein 2, is a 228 amino acid protein, which contains one THAP-type zinc finger. Members of the THAP (thanatos-associated protein) family of proteins contain a well conserved DNA-binding domain known as the THAP-type zinc finger motif. Proteins containing the THAP-type zinc finger motif are commonly involved in transcriptional regulation, cell-cycle control, apoptosis and chromatin modification

Western Blot: HEK-293 Cells, 1:200-1:2000; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: THAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR