Anti-CXorf22 Rabbit Polyclonal Antibody

Supplier: Proteintech
20837-1-AP-150UL
10085-444EA 402 USD
10085-444
Anti-CXorf22 Rabbit Polyclonal Antibody
Antibodies
IHC: Human lung Tissue, 1:20-1:200; Western Blot: Mouse lung Tissue, 1:200-1:2000

Type: Primary
Antigen: CXorf22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR