Anti-HDHD3 Rabbit Polyclonal Antibody

Supplier: Proteintech
20821-1-AP-150UL
10088-132EA 402 USD
10088-132
Anti-HDHD3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Colon Tissue, 1:500-1:5000

Type: Primary
Antigen: HDHD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR