Anti-C16orf14 Rabbit Polyclonal Antibody

Supplier: Proteintech
20808-1-AP-150UL
10083-780EA 402 USD
10083-780
Anti-C16orf14 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse skin Tissue, 1:1000-1:10000; IF: Hela, 1:10-1:100

Type: Primary
Antigen: C16orf14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR