Anti-neuron navigator 1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20807-1-AP-150UL
10090-836EA 402 USD
10090-836
Anti-neuron navigator 1 Rabbit Polyclonal Antibody
Antibodies
Neuron navigator 1 (NAV1) gene encodes a large protein belongs to the Nav/unc-53 family and 7 isoforms of NAV1 are produced by alternative splicing. NAV1 is localized in cytoplasm and expressed at high levels in heart, skeletal muscle and placenta, and especially in neuronal growth cones. Besides its nucleotide binding activity, NAV1 interacts with tubulin and associates with a subset of microtubule plus end, therefore playing a role in neuronal migration.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human Placenta Tissue, 1:20-1:200

Type: Primary
Antigen: neuron navigator 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR