Anti-ZNF385D Rabbit Polyclonal Antibody

Supplier: Proteintech
20806-1-AP-150UL
10097-344EA 402 USD
10097-344
Anti-ZNF385D Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Kidney Tissue, 1:200-1:2000

Type: Primary
Antigen: ZNF385D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR