Anti-C16orf13 Rabbit Polyclonal Antibody

Supplier: Proteintech
20801-1-AP-150UL
10083-778EA 402 USD
10083-778
Anti-C16orf13 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Liver Tissue, 1:500-1:5000

Type: Primary
Antigen: C16orf13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR