Anti-C7ORF50 Rabbit Polyclonal Antibody

Supplier: Proteintech
20797-1-AP-150UL
10083-890EA 402 USD
10083-890
Anti-C7ORF50 Rabbit Polyclonal Antibody
Antibodies
Western Blot:L02 Cells, 1:200-1:2000

Type: Primary
Antigen: C7ORF50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR