Anti-C11ORF70 Rabbit Polyclonal Antibody

Supplier: Proteintech
20780-1-AP-150UL
10083-756EA 402 USD
10083-756
Anti-C11ORF70 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human Testis, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: C11ORF70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR