Anti-HPDL Rabbit Polyclonal Antibody

Supplier: Proteintech
20777-1-AP-150UL
10088-426EA 402 USD
10088-426
Anti-HPDL Rabbit Polyclonal Antibody
Antibodies
Western Blot:L02 Cells, 1:500-1:5000; IHC: Human Stomach, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: HPDL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR