Anti-Serum amyloid P component Rabbit Polyclonal Antibody

Supplier: Proteintech
20773-1-AP-150UL
10082-988EA 402 USD
10082-988
Anti-Serum amyloid P component Rabbit Polyclonal Antibody
Antibodies
Serum amyloid P component (SAP), a member of pentraxin family, is a constitutive serum protein that is synthesized by hepatocytes. It is an acute phase protein, structurally related to C-reactive protein. SAP is a calcium-dependent ligand binding protein, which can interact with DNA and histones and may scavenge nuclear material released from damaged circulating cells.

Western Blot: Human Kidney Tissue, 1:500-1:5000; IHC: Human spleen Tissue, 1:20-1:200

Type: Primary
Antigen: Serum amyloid P component
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR