Anti-C14ORF133 Rabbit Polyclonal Antibody

Supplier: Proteintech
20771-1-AP-150UL
10083-762EA 402 USD
10083-762
Anti-C14ORF133 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:1000-1:10000; IF: Hela, 1:10-1:100

Type: Primary
Antigen: C14ORF133
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR