Anti-NPT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20751-1-AP-150UL
10090-906EA 402 USD
10090-906
Anti-NPT1 Rabbit Polyclonal Antibody
Antibodies
NPT1 (also known as SLC17A1) is a member of sodium-dependent inorganic phosphate transporters (NPT), which regulates entrance into the cellular membrane. NPT1 is mainly expressed in the kidney transporting small organic anions such as PAH (para-aminohippurate), but it is also found in the liver and brain. NTP1 localizes to the apical membrane of renal proximal tubular cells. NPT1 exits two isoforms due to the alternative splicing. Western blot analysis using this antibody detected a major band around 45-55kD in kidney tissue.

Western Blot: Human Placenta Tissue, 1:500-1:5000

Type: Primary
Antigen: NPT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR